Assay Details
| Gene Symbol: Tbc1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285991.1 | NP_001272920.1 | 16 - 17 | 1684 | 105 | |
| NM_001285993.1 | NP_001272922.1 | 16 - 17 | 1651 | 105 | ||
| NM_028162.4 | NP_082438.3 | 16 - 17 | 1684 | 105 | ||
| GenBank mRNA | AK144467.1 | - | 16 - 17 | 1685 | 105 | |
| AK154975.1 | - | 16 - 17 | 1644 | 105 | ||
| AK163366.1 | - | 16 - 17 | 1713 | 105 | ||
| AK220251.1 | - | 16 - 17 | 1651 | 105 | ||
| BC043113.1 | - | 16 - 17 | 1500 | 6407234 | 105 | |
| BC098328.1 | - | 11 - 12 | 1030 | 30357143 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d5
Genomic Map