Assay Details
| Gene Symbol: Mcph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173189.2 | NP_775281.2 | 4 - 5 | 764 | 142 | |
| GenBank mRNA | AK083572.1 | - | 4 - 5 | 807 | 142 | |
| AK147997.1 | - | 4 - 5 | 356 | 142 | ||
| AK148021.1 | - | 4 - 5 | 362 | 142 | ||
| AY070216.1 | - | 4 - 5 | 344 | 142 | ||
| BC056924.1 | - | 4 - 5 | 764 | 5697978 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mcph1
Genomic Map