Assay Details
| Gene Symbol: Sgsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197943.2 | NP_922934.2 | 20 - 21 | 2864 | 117 | |
| GenBank mRNA | AK122273.1 | - | 20 - 21 | 2786 | 117 | |
| AK199338.1 | - | 2 - 3 | 164 | 117 | ||
| AK199845.1 | - | 2 - 3 | 164 | 117 | ||
| AK203050.1 | - | 2 - 3 | 164 | 117 | ||
| AK212740.1 | - | 2 - 3 | 164 | 117 | ||
| AK213455.1 | - | 2 - 3 | 164 | 117 | ||
| BC058414.1 | - | 20 - 21 | 2855 | 6822416 | 117 | |
| BC060163.1 | - | 20 - 21 | 2855 | 6822416 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm2
Genomic Map