Assay Details
| Gene Symbol: Adipor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197985.3 | NP_932102.2 | 1 - 2 | 66 | 108 | |
| GenBank mRNA | AK039667.1 | - | 1 - 2 | 274 | 108 | |
| AK046591.1 | - | 1 - 2 | 73 | 108 | ||
| AK134787.1 | - | 1 - 2 | 82 | 108 | ||
| AK160534.1 | - | 1 - 2 | 83 | 108 | ||
| AY424291.1 | - | 1 - 2 | 66 | 108 | ||
| BC024094.1 | - | 1 - 2 | 66 | 5132696 | 108 | |
| BC064109.1 | - | 1 - 2 | 64 | 3371697 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK039667.1, AK046591.1, AK134787.1, AK160534.1, AY424291.1, BC024094.1, BC064109.1, NM_197985.3 transcripts.
Set Membership: |
|

Adipor2
Genomic Map