Assay Details
| Gene Symbol: Hsh2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197944.1 | NP_922935.1 | 6 | 1025 | 88 | |
| GenBank mRNA | AK157000.1 | - | 1058 | 88 | ||
| AY319653.1 | - | 6 | 1025 | 88 | ||
| BC094291.1 | - | 5 | 1004 | 5316178 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522427 | BC094291.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522427 | NM_197944.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522427 | AK157000.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522427 | AY319653.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522427 | BC094291.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522427 | NM_197944.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522427 | AK157000.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522427 | AY319653.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497693 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465117 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572789 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 363181 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50640 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161888 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101761 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522427 | BC094291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522427 | NM_197944.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522427 | AK157000.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522427 | AY319653.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522427 | BC094291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522427 | NM_197944.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522427 | AK157000.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522427 | AY319653.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25960091 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101761 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01765256 | - | - |
| Whole Mouse Genome G4122A | A_52_P522427 | BC094291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522427 | NM_197944.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522427 | AK157000.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522427 | AY319653.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522427 | BC094291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522427 | NM_197944.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522427 | AK157000.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522427 | AY319653.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136328 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101761 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06926785 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06926780 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06926783 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06727564 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06727565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25960204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07445474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07118227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07118226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06691061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07306759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07355693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06715997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07320104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06954573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06715247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07198132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07440109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06758911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07440104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07420243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07101670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25960098 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10572777 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572776 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572774 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572773 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572775 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442130_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10572772 | - | - |
| Mouse Genome 430 2.0 Array | 1442130_at | - | - |
| Mouse Genome 430 2.0 Array | 1442130_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800006012.mm.1 | TC0800000817.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1442130_PM_at | - | - |
| Mouse Expression Set 430 | 1442130_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442130_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442130_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 840106 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2060494 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002060494 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239119 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239119 | - | - |
| mouse-6 v1.1 (Illumina) | scl0209488.6_203-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2060494 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0209488.6_203-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2060494 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239119 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239119 | - | - |

Hsh2d
Genomic Map