Assay Details
| Gene Symbol: Ssh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198113.2 | NP_932781.1 | 1 - 2 | 153 | 68 | |
| GenBank mRNA | AB099289.1 | - | 1 - 2 | 151 | 68 | |
| AK148293.1 | - | 1 - 2 | 147 | 68 | ||
| AK149880.1 | - | 1 - 2 | 162 | 68 | ||
| AK164662.1 | - | 1 - 2 | 83 | 68 | ||
| AK171514.1 | - | 1 - 2 | 150 | 68 | ||
| BC028922.1 | - | 1 - 2 | 153 | 3987714 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ssh3
Genomic Map