Assay Details
| Gene Symbol: Repin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175099.3 | NP_780308.2 | 1 - 2 | 169 | 88 | |
| GenBank mRNA | AK089041.1 | - | 1 - 2 | 113 | 88 | |
| BM943840.1 | - | 1 - 2 | 127 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089041.1, NM_175099.3 transcripts.
Set Membership: |
|

Repin1
Genomic Map