Assay Details
| Gene Symbol: Cnga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007723.2 | NP_031749.2 | 7 - 8 | 795 | 148 | |
| GenBank mRNA | BC141261.1 | - | 7 - 8 | 564 | 9055873 | 148 |
| CO423854.1 | - | 2 - 3 | 174 | 148 | ||
| M84742.1 | - | 7 - 8 | 564 | 148 | ||
| U19717.1 | - | 6 - 7 | 530 | 148 | ||
More Information
Set Membership: |
|

Cnga1
Genomic Map