Assay Details
| Gene Symbol: Krt84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008474.2 | NP_032500.2 | 8 - 9 | 1549 | 147 | |
| GenBank mRNA | AY028607.1 | - | 8 - 9 | 1506 | 147 | |
| BC114971.1 | - | 8 - 9 | 1502 | 40039508 | 147 | |
| X65505.1 | - | 5 - 6 | 569 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145601 | X65505.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145601 | NM_008474.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145601 | AA738772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145601 | AY028607.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109772 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145601 | X65505.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145601 | NM_008474.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145601 | AA738772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145601 | AY028607.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P367880 | X65505.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P367880 | NM_008474.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P367880 | AA738772.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P367880 | AY028607.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30492481 | - | - |
| Whole Mouse Genome G4122A | A_51_P367880 | X65505.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P367880 | NM_008474.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P367880 | AA738772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P367880 | AY028607.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106901 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137974 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12385062 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12385065 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12385045 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12385033 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12385036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30492514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12102349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12294217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12228360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12385063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12385049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11845286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11845280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12102350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12228365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12385029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11934935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11934920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11934939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11845288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12228364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11942242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12385060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30492467 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10432717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432712 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432718 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432713 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451551_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10432711 | - | - |
| Mouse Genome 430 2.0 Array | 1451551_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451551_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16680_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16680_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500018586.mm.1 | NM_008474.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018586.mm.1 | NM_008474.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018586.mm.1 | TC1500002261.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018587.mm.1 | NM_008474.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018587.mm.1 | NM_008474.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018587.mm.1 | TC1500002261.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451551_PM_at | - | - |
| Mouse Expression Set 430 | 1451551_at | - | - |
| Murine Genome U74v2 Set | 96503_PM_at | - | - |
| Murine Genome U74v2 Set | 96503_at | - | - |
| Murine Genome U74v2 Set | 162328_f_at | - | - |
| Murine Genome U74v2 Set | 162328_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451551_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451551_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451551_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451551_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 745256 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6980086 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6980086 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006980086 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2739740 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2739740 | - | - |
| mouse-6 v1.1 (Illumina) | scl46708.9.1_81-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6980086 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46708.9.1_81-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980086 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2739740 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2739740 | - | - |

Krt84
Genomic Map