Assay Details
| Gene Symbol: Serpina1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009247.2 | NP_033273.1 | 2 - 3 | 672 | 131 | |
| GenBank mRNA | AK004999.1 | - | 2 - 3 | 674 | 131 | |
| AK146576.1 | - | 874 | 131 | |||
| BC061176.1 | - | 2 - 3 | 683 | 30381238 | 131 | |
| M75717.1 | - | 2 - 3 | 667 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of peptidase activity
negative regulation of endopeptidase activity
response to cytokine
response to peptide hormone
glycoprotein binding
protease binding
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
peptidase inhibitor activity
identical protein binding
negative regulation of peptidase activity
negative regulation of endopeptidase activity
response to cytokine
response to peptide hormone
glycoprotein binding
protease binding
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
peptidase inhibitor activity
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010292 | AK146576.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010292 | BC061176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010292 | M75717.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010292 | AK004999.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010292 | NM_009247.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113857 | AK146576.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113857 | BC061176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113857 | NM_009247.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113857 | AK135650.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630457 | AK036338.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630457 | AK084510.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630457 | AK089082.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630457 | AK162446.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630457 | AK214002.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630457 | AY007813.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630457 | BC018326.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630457 | NM_030705.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 20433 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113857 | AK146576.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113857 | BC061176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113857 | NM_009247.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113857 | AK135650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010292 | AK146576.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010292 | BC061176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010292 | M75717.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010292 | AK004999.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010292 | NM_009247.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28700527 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07733367 | - | - |
| Whole Mouse Genome G4122A | A_52_P598495 | AK146576.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598495 | BC061176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598495 | M75717.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598495 | AK004999.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P598495 | M75718.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28700562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09870028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09870026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10176373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09780476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09856995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09823424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09856991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09815174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09870030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28700556 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10402414 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402411 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402412 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402413 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402402 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402410 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10402399 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10402409 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200015906.mm.1 | NM_009247.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015906.mm.1 | NM_009247.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015906.mm.1 | TC1200002285.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200015907.mm.1 | NM_009247.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015907.mm.1 | NM_009247.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015907.mm.1 | TC1200002285.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200015905.mm.1 | NM_009247.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015905.mm.1 | NM_009247.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200015905.mm.1 | TC1200002285.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449321_PM_x_at | - | - |
| Mouse Expression Set 430 | 1449321_x_at | - | - |
| Murine Genome U74v2 Set | 101574_PM_f_at | - | - |
| Murine Genome U74v2 Set | 101574_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449321_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449321_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 389786 | - | - |
| ABI Mouse Genome Survey Microarray | 610707 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2630180 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2630180 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002630180 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220234 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220234 | - | - |
| mouse-6 v1.1 (Illumina) | scl0020704.1_175-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2630180 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0020704.1_175-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630180 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1220234 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220234 | - | - |

Serpina1e
Genomic Map