Assay Details
| Gene Symbol: Rab5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177411.4 | NP_803130.1 | 2 - 3 | 345 | 99 | |
| GenBank mRNA | AB232594.1 | - | 1 - 2 | 206 | 99 | |
| AK081251.1 | - | 2 - 3 | 341 | 99 | ||
| AK083034.1 | - | 2 - 3 | 334 | 99 | ||
| AK144551.1 | - | 2 - 3 | 467 | 99 | ||
| BC137574.1 | - | 1 - 2 | 257 | 8860593 | 99 | |
| BC137575.1 | - | 1 - 2 | 257 | 8860594 | 99 | |
| BY727964.1 | - | 2 - 3 | 346 | 99 | ||
| X84239.1 | - | 1 - 2 | 214 | 99 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Delineating biological pathways unique to embryonic stem cell-derived insulin-producing cell lines from their noninsulin-producing progenitor cell lines.
- Authors
- Chen TS, Tan SS, Yeo RW, Teh BJ, Luo R, Li G, Lim SK
- Journal
- Endocrinology - Page No 3600 (Sun Aug 01 00:00:00 EDT 2010)
- Volume (Year)
- 151 (2010)
- Product Usage
- -

Rab5b
Genomic Map