Assay Details
| Gene Symbol: Edf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021519.1 | NP_067494.1 | 2 - 3 | 182 | 106 | |
| GenBank mRNA | AB030185.1 | - | 2 - 3 | 182 | 106 | |
| AJ309569.1 | - | 2 - 3 | 130 | 106 | ||
| AK002345.1 | - | 2 - 3 | 197 | 106 | ||
| AK003557.1 | - | 2 - 3 | 209 | 106 | ||
| AK010191.1 | - | 2 - 3 | 197 | 106 | ||
| BC023472.1 | - | 2 - 3 | 177 | 5250117 | 106 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
cell differentiation
positive regulation of DNA binding
regulation of nucleic acid-templated transcription
DNA binding
transcription coactivator activity
calmodulin binding
sequence-specific DNA binding
poly(A) RNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
cell differentiation
positive regulation of DNA binding
regulation of nucleic acid-templated transcription
DNA binding
transcription coactivator activity
calmodulin binding
sequence-specific DNA binding
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21072938 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21072941 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21072937 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21072940 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007535 | NM_021519.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007535 | AB030185.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574231 | AK133982.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574231 | BC060147.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574231 | NM_207234.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45972 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102439 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1992388 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007535 | NM_021519.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007535 | AB030185.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160567 | BC023472.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160567 | NM_021519.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160567 | AB030185.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160567 | AJ309569.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160567 | AK002345.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21072933 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102439 | - | - |
| Whole Mouse Genome G4122A | A_51_P160567 | BC023472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160567 | NM_021519.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160567 | AB030185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160567 | AJ309569.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160567 | AK002345.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101908 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102439 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01741332 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01741331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21072940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21072937 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21072941 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21072938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21072917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01300609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01237848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01300607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01933418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01237851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01558922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01237858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02212736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01300614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01801576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02212738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01300618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01933417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21072955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01300611 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10470121 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470124 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470122 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470120 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470123 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438503_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440151_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448266_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10470119 | - | - |
| Mouse Genome 430 2.0 Array | 1440152_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1440152_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1440151_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1440151_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1438503_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1448266_at | - | - |
| Mouse Genome 430 2.0 Array | 1438503_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1448266_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 59022_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 59022_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200002562.mm.1 | NM_021519.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002562.mm.1 | NM_021519.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002562.mm.1 | TC0200000422.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200002559.mm.1 | NM_021519.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002559.mm.1 | NM_021519.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002559.mm.1 | TC0200000422.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200002558.mm.1 | NM_021519.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002558.mm.1 | NM_021519.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002558.mm.1 | TC0200000422.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200002566.mm.1 | NM_021519.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002566.mm.1 | NM_021519.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002566.mm.1 | TC0200000422.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200002560.mm.1 | NM_021519.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002560.mm.1 | NM_021519.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002560.mm.1 | TC0200000422.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200002556.mm.1 | NM_021519.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002556.mm.1 | NM_021519.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002556.mm.1 | TC0200000422.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438503_x_at | - | - |
| Mouse Expression Set 430 | 1438503_PM_x_at | - | - |
| Mouse Expression Set 430 | 1448266_at | - | - |
| Mouse Expression Set 430 | 1448266_PM_at | - | - |
| Mouse Expression Set 430 | 1440152_PM_x_at | - | - |
| Mouse Expression Set 430 | 1440152_x_at | - | - |
| Mouse Expression Set 430 | 1440151_s_at | - | - |
| Mouse Expression Set 430 | 1440151_PM_s_at | - | - |
| Murine Genome U74v2 Set | 96322_at | - | - |
| Murine Genome U74v2 Set | 96322_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438503_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448266_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438503_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448266_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448266_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448266_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440151_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440151_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 900995 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1660035 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2640161 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1660035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2640161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001660035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002640161 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2739033 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2739033 | - | - |
| mouse-6 v1.1 (Illumina) | scl059022.3_20-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003286.1_154-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1660035 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2640161 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003286.1_154-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl059022.3_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640161 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2739033 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2739033 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2662831 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl059022.3_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003286.1_154-S | - | - |

Edf1
Genomic Map