Assay Details
| Gene Symbol: Trmt112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166370.1 | NP_001159842.1 | 2 - 3 | 667 | 62 | |
| NM_026306.3 | NP_080582.3 | 3 - 4 | 569 | 62 | ||
| GenBank mRNA | AK002791.1 | - | 2 - 3 | 214 | 62 | |
| AK003292.1 | - | 2 - 3 | 212 | 62 | ||
| BC016191.1 | - | 2 - 3 | 589 | 4483787 | 62 | |
| BC019418.1 | - | 2 - 3 | 201 | 4162006 | 62 | |
| BC087959.1 | - | 3 - 4 | 569 | 6766768 | 62 | |
| BE951374.1 | - | 2 - 3 | 62 | |||
| DV654053.1 | - | 2 - 3 | 557 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Trmt112
Genomic Map