Assay Details
| Gene Symbol: Rpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026632.4 | NP_080908.1 | 2 - 3 | 221 | 77 | |
| GenBank mRNA | AK005285.1 | - | 2 - 3 | 194 | 77 | |
| AK009916.1 | - | 2 - 3 | 193 | 77 | ||
| AK021228.1 | - | 2 - 3 | 194 | 77 | ||
| BC028489.1 | - | 2 - 3 | 186 | 1448430 | 77 | |
| BY057686.1 | - | 2 - 3 | 222 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpa3
Genomic Map