Assay Details
| Gene Symbol: Tmem50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027935.2 | NP_082211.1 | 3 - 4 | 244 | 61 | |
| GenBank mRNA | AK014282.1 | - | 3 - 4 | 314 | 61 | |
| AK147069.1 | - | 3 - 4 | 280 | 61 | ||
| AK152630.1 | - | 3 - 4 | 328 | 61 | ||
| AK152654.1 | - | 3 - 4 | 299 | 61 | ||
| AK157214.1 | - | 3 - 4 | 317 | 61 | ||
| AK164989.1 | - | 3 - 4 | 310 | 61 | ||
| AK165636.1 | - | 3 - 4 | 293 | 61 | ||
| BC094256.1 | - | 3 - 4 | 244 | 6820666 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem50a
Genomic Map