Assay Details
| Gene Symbol: Tmem50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027935.2 | NP_082211.1 | 5 - 6 | 404 | 68 | |
| GenBank mRNA | AK014282.1 | - | 5 - 6 | 474 | 68 | |
| AK147069.1 | - | 5 - 6 | 440 | 68 | ||
| AK152654.1 | - | 5 - 6 | 459 | 68 | ||
| AK157214.1 | - | 5 - 6 | 477 | 68 | ||
| AK164989.1 | - | 5 - 6 | 470 | 68 | ||
| AK165636.1 | - | 5 - 6 | 453 | 68 | ||
| BC094256.1 | - | 5 - 6 | 404 | 6820666 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem50a
Genomic Map