Assay Details
| Gene Symbol: Mrpl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053161.2 | NP_444391.1 | 3 - 4 | 249 | 153 | |
| GenBank mRNA | AB049648.1 | - | 3 - 4 | 244 | 153 | |
| AK143575.1 | - | 3 - 4 | 277 | 154 | ||
| BC030074.1 | - | 3 - 4 | 249 | 1514322 | 153 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28034150 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2073855 | AK143575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2073855 | BC030074.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2073855 | BC096568.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2073855 | NM_053161.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104810 | AB049648.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104810 | AK143575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104810 | BC030074.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104810 | BC096568.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104810 | NM_053161.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333460 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420899 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42416 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176174 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99416 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180346 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2073855 | AK143575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2073855 | BC030074.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2073855 | BC096568.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2073855 | NM_053161.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104810 | AB049648.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104810 | AK143575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104810 | BC030074.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104810 | BC096568.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104810 | NM_053161.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272175 | AB049648.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272175 | AK143575.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272175 | BC030074.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272175 | BC096568.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P272175 | NM_053161.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28034170 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180346 | - | - |
| Whole Mouse Genome G4122A | A_51_P272175 | AB049648.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272175 | AK143575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272175 | BC030074.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272175 | BC096568.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272175 | NM_053161.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138041 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130351 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180346 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09261828 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09261827 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09537694 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28034150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28034248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09039343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09261828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09039341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09537692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09140801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28034187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09183036 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10454094 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380414 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380412 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380413 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415690_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10454093 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380411 | - | - |
| Mouse Genome 430 2.0 Array | 1415690_at | - | - |
| Mouse Genome 430 2.0 Array | 1415690_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94064_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94064_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100013386.mm.1 | NM_053161.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013386.mm.1 | NM_053161.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013386.mm.1 | TC1100001443.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013387.mm.1 | NM_053161.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013387.mm.1 | NM_053161.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013387.mm.1 | TC1100001443.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013391.mm.1 | NM_053161.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013391.mm.1 | NM_053161.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013391.mm.1 | TC1100001443.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013380.mm.1 | NM_053161.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013380.mm.1 | NM_053161.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013380.mm.1 | TC1100001443.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013390.mm.1 | NM_053161.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013390.mm.1 | NM_053161.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013390.mm.1 | TC1100001443.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100013389.mm.1 | NM_053161.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013389.mm.1 | NM_053161.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100013389.mm.1 | TC1100001443.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1415690_PM_at | - | - |
| Mouse Expression Set 430 | 1415690_at | - | - |
| Mouse Expression Set 430 | 1442299_at | - | - |
| Mouse Expression Set 430 | 1442299_PM_at | - | - |
| Mouse Expression Set 430 | 1415690_PM_at | - | - |
| Mouse Expression Set 430 | 1415690_at | - | - |
| Murine Genome U74v2 Set | 98120_at | - | - |
| Murine Genome U74v2 Set | 98120_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415690_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415690_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101190075 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2060131 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101190075 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002060131 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2691298 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2691298 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256266 | - | - |
| mouse-6 v1.1 (Illumina) | scl094064.4_160-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001301.1_12-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101190075 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2060131 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl094064.4_160-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2060131 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101190075 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2691298 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2691298 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl094064.4_160-S | - | - |

Mrpl27
Genomic Map