Assay Details
Gene Symbol: Vmn1r43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053220.2 | NP_444450.2 | 1 | 720 | 167 | |
GenBank mRNA | BC125364.1 | - | 1 | 720 | 40129879 | 167 |
BC132284.1 | - | 1 | 720 | 40130561 | 167 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055717 | BC125364.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055717 | NM_053220.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055717 | BC132284.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055717 | BC125364.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055717 | NM_053220.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055717 | BC132284.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P409868 | BC125364.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P409868 | NM_053220.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P409868 | BC132284.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630123 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630136 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630141 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630166 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630157 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630147 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630151 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630149 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630167 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630171 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630169 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630138 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630126 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630162 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630164 | - | - |
Whole Mouse Genome G4122A | A_51_P409868 | BC125364.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P409868 | NM_053220.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P409868 | BC132284.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128268 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05812463 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05812457 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05812479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630136 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05063265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05595846 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630149 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630136 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630160 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05063253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630153 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05063266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630169 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630138 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05874232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630147 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05063263 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05812475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05812479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630145 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10546234 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422368_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10546233 | - | - |
Mouse Genome 430 2.0 Array | 1422368_at | - | - |
Mouse Genome 430 2.0 Array | 1422368_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 113847_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 113847_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600021051.mm.1 | TC0600002705.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600021051.mm.1 | NM_053220.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600021051.mm.1 | NM_053220.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600021052.mm.1 | TC0600002705.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600021052.mm.1 | NM_053220.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600021052.mm.1 | NM_053220.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600021050.mm.1 | TC0600002705.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600021050.mm.1 | NM_053220.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600021050.mm.1 | NM_053220.2 | Refseq |
Mouse Expression Set 430 | 1422368_at | - | - |
Mouse Expression Set 430 | 1422368_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422368_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422368_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422368_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422368_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 390817 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161596 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3161596 | - | - |
mouse-6 v1.1 (Illumina) | GI_16716526-S | - | - |
