Assay Details
| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145400.3 | NP_663375.3 | 9 - 10 | 1361 | 153 | |
| GenBank mRNA | AK172893.1 | - | 9 - 10 | 1321 | 153 | |
| BC006649.1 | - | 10 - 11 | 1500 | 3498834 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube4a
Genomic Map