Assay Details
Gene Symbol: Olfr1330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146334.2 | NP_666446.1 | 1 | 754 | 198 | |
GenBank mRNA | BC108943.1 | - | 1 | 754 | 40042936 | 198 |
BC108944.1 | - | 1 | 749 | 198 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2158023 | BC108943.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2158023 | BC108944.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2158023 | NM_146334.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2158023 | BC108943.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2158023 | BC108944.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2158023 | NM_146334.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P313729 | BC108943.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P313729 | BC108944.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P313729 | NM_146334.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242606 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242639 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242605 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242604 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242623 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242611 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242603 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242647 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242601 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242619 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242612 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242638 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242642 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242629 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242640 | - | - |
Whole Mouse Genome G4122A | A_51_P313729 | BC108943.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P313729 | BC108944.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P313729 | NM_146334.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102127 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242611 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242623 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242606 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242598 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242603 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242611 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242638 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242604 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242593 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242601 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242639 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242610 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242654 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242615 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03942745 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03999615 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03372901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242629 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242597 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242652 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242640 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242648 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242622 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03629193 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242642 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242630 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10515827 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10507577 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515831 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10507579 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10515826 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10507578 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 893302 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5050300 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005050300 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250973 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1250973 | - | - |
mouse-6 v1.1 (Illumina) | scl3022.1.1_75-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5050300 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl3022.1.1_75-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5050300 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2864972 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1250973 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250973 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2864972 | - | - |
