Assay Details
Gene Symbol: Olfr164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146451.1 | NP_666662.1 | 1 | 445 | 174 | |
GenBank mRNA | BC145676.1 | - | 1 | 541 | 40130979 | 174 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005512 | BC145676.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005512 | NM_146451.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122787 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005512 | BC145676.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005512 | NM_146451.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P308697 | BC145676.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P308697 | NM_146451.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583588 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583585 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583577 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583572 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583574 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583569 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583581 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583589 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583570 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583587 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583595 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583580 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583575 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30583593 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122787 | - | - |
Whole Mouse Genome G4122A | A_51_P308697 | BC145676.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P308697 | NM_146451.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131696 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122787 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13050903 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12692024 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12692029 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12791542 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13050899 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12692034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30583587 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583554 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12692031 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583586 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583593 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583587 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583572 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583557 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583565 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12692040 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583541 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583576 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583592 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583589 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583544 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583560 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583552 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583543 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583599 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12692024 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12483436 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583555 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583563 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583542 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583550 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583595 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583581 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583580 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583588 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583573 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30583547 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10438422 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10438421 | - | - |
Affymetrix Mouse Clariom S Array | PSR1600010977.mm.1 | TC1600001327.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600010976.mm.1 | NM_146451.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600010976.mm.1 | NM_146451.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600010976.mm.1 | TC1600001327.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 339437 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 730435 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 730435 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000730435 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2719434 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2719434 | - | - |
mouse-6 v1.1 (Illumina) | scl601.1.1_67-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 730435 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl601.1.1_67-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 730435 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2839732 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2839732 | - | - |
