Assay Details
| Gene Symbol: Olfr98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146510.1 | NP_666721.1 | 1 | 845 | 152 | |
| GenBank mRNA | BC119060.1 | - | 1 | 932 | 8733809 | 152 |
| BC120744.1 | - | 1 | 932 | 40129667 | 152 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63623 | NM_146510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63623 | BC119060.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63623 | BC120744.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63623 | NM_146510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63623 | BC119060.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63623 | BC120744.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131470 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63623 | NM_146510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63623 | BC119060.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63623 | BC120744.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63623 | NM_146510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63623 | BC119060.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63623 | BC120744.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P63623 | NM_146510.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P63623 | BC119060.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P63623 | BC120744.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P63623 | NM_146510.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P63623 | BC119060.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P63623 | BC120744.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168074 | - | - |
| Whole Mouse Genome G4122A | A_52_P63623 | NM_146510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63623 | BC119060.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63623 | BC120744.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63623 | NM_146510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63623 | BC119060.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63623 | BC120744.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110727 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140843 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13344523 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13091083 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13130101 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13091096 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13344526 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13373550 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13373553 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13130096 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13130097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13207339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13593213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13207343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13130096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13513159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31168068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13207342 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450869 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450868 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700019553.mm.1 | NM_146510.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019553.mm.1 | NM_146510.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019553.mm.1 | TC1700002061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019555.mm.1 | TC1700002061.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 897469 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2900500 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2900500 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002900500 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2589485 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2589485 | - | - |
| mouse-6 v1.1 (Illumina) | scl49932.1.1_31-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2900500 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49932.1.1_31-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900500 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2589485 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2589485 | - | - |

Olfr98
Genomic Map