Assay Details
| Gene Symbol: Olfr854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146522.1 | NP_666733.1 | 1 | 237 | 143 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964413 | NM_146522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964413 | NM_146522.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160692 | NM_146522.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340305 | - | - |
| Whole Mouse Genome G4122A | A_51_P160692 | NM_146522.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135725 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07682864 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07620369 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07682867 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07682865 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07620365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08127125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340273 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591199 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591198 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 794360 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6040524 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040524 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2665527 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2665527 | - | - |
| mouse-6 v1.1 (Illumina) | scl36176.1.337_35-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040524 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36176.1.337_35-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040524 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2665527 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2665527 | - | - |

Olfr854
Genomic Map