Assay Details
Gene Symbol: Olfr822 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146671.1 | NP_666882.1 | 1 | 682 | 162 | |
GenBank mRNA | BC127972.1 | - | 1 | 686 | 40135122 | 162 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406204 | BC127972.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406204 | NM_146671.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406204 | BC127972.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406204 | NM_146671.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66839 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144191 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406204 | BC127972.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406204 | NM_146671.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406204 | BC127972.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406204 | NM_146671.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P406204 | BC127972.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P406204 | NM_146671.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P406204 | BC127972.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P406204 | NM_146671.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548269 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548239 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548241 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548220 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548232 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548235 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548270 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548225 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548242 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548238 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548222 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548221 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548224 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548229 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548256 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548234 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548246 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27548233 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144191 | - | - |
Whole Mouse Genome G4122A | A_51_P406204 | BC127972.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406204 | NM_146671.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406204 | BC127972.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P406204 | NM_146671.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136018 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144191 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08459366 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08459370 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08459374 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08565408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27548229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548238 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548225 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548261 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08569378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548251 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548246 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548241 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548270 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27548221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08565407 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08730197 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10367531 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10367530 | - | - |
Affymetrix Mouse Clariom S Array | PSR1000012062.mm.1 | NM_146671.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012062.mm.1 | NM_146671.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012062.mm.1 | TC1000001659.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 818902 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4060538 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004060538 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1246505 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1246505 | - | - |
mouse-6 v1.1 (Illumina) | scl0258666.1_1-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4060538 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258666.1_1-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060538 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1246505 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1246505 | - | - |
