Assay Details
Gene Symbol: Olfr912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146810.2 | NP_667021.2 | 2 | 131 | 153 | |
GenBank mRNA | CB174492.1 | - | 2 | 131 | 153 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201400 | NM_146810.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201400 | CB174340.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201400 | NM_146810.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201400 | CB174340.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201400 | NM_146810.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201400 | CB174340.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201400 | NM_146810.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201400 | CB174340.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P201400 | NM_146810.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P201400 | CB174340.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P201400 | NM_146810.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P201400 | CB174340.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435209 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435216 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435242 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435225 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435233 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435229 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435246 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435222 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435227 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435230 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435212 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435232 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435220 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435211 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435218 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435235 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435215 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435214 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435224 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435247 | - | - |
Whole Mouse Genome G4122A | A_51_P201400 | NM_146810.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P201400 | CB174340.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P201400 | NM_146810.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P201400 | CB174340.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119232 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131812 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120418 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07791276 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07791275 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07737978 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07737976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435209 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435227 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435246 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435236 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435254 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435230 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435233 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08008714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07791277 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435245 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07737976 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07457713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435225 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435215 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435218 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435211 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435242 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10584429 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10584430 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10584428 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3130167 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1246906 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1246906 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2739404 | - | - |
