Assay Details
| Gene Symbol: Olfr45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146963.1 | NP_667174.1 | 1 | 446 | 237 | |
| GenBank mRNA | AF102519.1 | - | 1 | 254 | 237 | |
| BC125298.1 | - | 1 | 469 | 40129813 | 237 | |
| BC125300.1 | - | 1 | 469 | 40129815 | 237 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428102 | AF102519.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428102 | BC125298.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428102 | BC125300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428102 | NM_146963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428102 | AF102519.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428102 | BC125298.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428102 | BC125300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P428102 | NM_146963.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172531 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428102 | AF102519.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428102 | BC125298.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428102 | BC125300.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428102 | NM_146963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428102 | AF102519.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428102 | BC125298.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428102 | BC125300.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P428102 | NM_146963.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428102 | AF102519.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428102 | BC125298.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428102 | BC125300.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428102 | NM_146963.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428102 | AF102519.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428102 | BC125298.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428102 | BC125300.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428102 | NM_146963.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25584487 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172531 | - | - |
| Whole Mouse Genome G4122A | A_51_P428102 | AF102519.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428102 | BC125298.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428102 | BC125300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428102 | NM_146963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428102 | AF102519.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428102 | BC125298.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428102 | BC125300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428102 | NM_146963.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130457 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06000606 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06000636 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06000625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25584502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06509362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06509361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06000636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06000611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25584482 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10558669 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558658 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10558668 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700017694.mm.1 | TC0700002020.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017694.mm.1 | NM_146963.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017694.mm.1 | NM_146963.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 484166 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 770068 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2603159 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2603159 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2603159 | - | - |

Olfr45
Genomic Map