Assay Details
| Gene Symbol: Myh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK081482.1 | - | 5 - 6 | 605 | 247 | |
| AK087387.1 | - | 5 - 6 | 605 | 247 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Myh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK081482.1 | - | 5 - 6 | 605 | 247 | |
| AK087387.1 | - | 5 - 6 | 605 | 247 | ||
Set Membership: |
|