Assay Details
| Gene Symbol: Tnfsf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183391.3 | NP_899247.3 | 2 - 3 | 220 | 71 | |
| GenBank mRNA | AJ577579.1 | - | 2 - 3 | 222 | 71 | |
| AJ577580.1 | - | 222 | 71 | |||
| AY234223.1 | - | 2 - 3 | 178 | 71 | ||
| AY320040.1 | - | 2 - 3 | 170 | 71 | ||
| AY359852.1 | - | 2 - 3 | 164 | 71 | ||
| BC137814.1 | - | 2 - 3 | 220 | 8860833 | 71 | |
| BC137815.1 | - | 2 - 3 | 220 | 8860834 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
T cell proliferation involved in immune response
immune system process
positive regulation of leukocyte migration
immune response
cell-cell signaling
positive regulation of macrophage chemotaxis
interleukin-10 production
interleukin-12 production
regulation of interferon-gamma production
regulation of interleukin-6 production
regulation of tumor necrosis factor production
tumor necrosis factor-mediated signaling pathway
activated CD4-positive, alpha-beta T cell proliferation
regulation of T cell proliferation
positive regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of cell adhesion
positive regulation of inflammatory response
positive regulation of NF-kappaB transcription factor activity
regulation of monocyte chemotactic protein-1 production
positive regulation of monocyte chemotaxis
positive regulation of protein homodimerization activity
negative regulation of T-helper 17 cell lineage commitment
regulation of CD40 signaling pathway
regulation of dendritic cell chemotaxis
cytokine activity
tumor necrosis factor receptor binding
protein binding
tumor necrosis factor receptor superfamily binding
T cell proliferation involved in immune response
immune system process
positive regulation of leukocyte migration
immune response
cell-cell signaling
positive regulation of macrophage chemotaxis
interleukin-10 production
interleukin-12 production
regulation of interferon-gamma production
regulation of interleukin-6 production
regulation of tumor necrosis factor production
tumor necrosis factor-mediated signaling pathway
activated CD4-positive, alpha-beta T cell proliferation
regulation of T cell proliferation
positive regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of cell adhesion
positive regulation of inflammatory response
positive regulation of NF-kappaB transcription factor activity
regulation of monocyte chemotactic protein-1 production
positive regulation of monocyte chemotaxis
positive regulation of protein homodimerization activity
negative regulation of T-helper 17 cell lineage commitment
regulation of CD40 signaling pathway
regulation of dendritic cell chemotaxis
cytokine activity
tumor necrosis factor receptor binding
protein binding
tumor necrosis factor receptor superfamily binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075553 | NM_183391.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075553 | AJ577579.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921104 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195875 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179839 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282767 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29492 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125728 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180518 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075553 | NM_183391.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075553 | AJ577579.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20772139 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180518 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125728 | - | - |
| Whole Mouse Genome G4122A | A_52_P603111 | NM_183391.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P603111 | AJ577579.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111657 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180518 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125728 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00043890 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00043896 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00780990 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00043888 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00043899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20772084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00856371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00856382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00607931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00043895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00834280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00043890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00603507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00341699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00341697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00781591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00791407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00227790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00791408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00043896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20772126 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351106 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351105 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351099 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100012100.mm.1 | NM_183391.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012100.mm.1 | NM_183391.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012100.mm.1 | TC0100001469.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100012101.mm.1 | NM_183391.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012101.mm.1 | NM_183391.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012101.mm.1 | TC0100001469.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 680388 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2850520 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1990458 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2850520 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002850520 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001990458 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2483100 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2482910 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2482910 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2483100 | - | - |
| mouse-6 v1.1 (Illumina) | scl00240873.2_20-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0240873.3_32-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2850520 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1990458 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00240873.2_20-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0240873.3_32-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990458 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850520 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2482910 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2482910 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0240873.3_32-S | - | - |

Tnfsf18
Genomic Map