Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400018466.mm.1 |
NM_194055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018466.mm.1 |
TC0400002208.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018465.mm.1 |
NM_194055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018465.mm.1 |
TC0400002208.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018463.mm.1 |
TC0400002208.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018464.mm.1 |
NM_194055.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018464.mm.1 |
TC0400002208.mm.1 |
MTA TC |