Assay Details
| Gene Symbol: Aldh3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112725.1 | NP_001106196.1 | 7 - 8 | 998 | 87 | |
| NM_007436.2 | NP_031462.2 | 7 - 8 | 1130 | 87 | ||
| GenBank mRNA | EL959139.1 | - | 2 - 3 | 277 | 87 | |
| U12785.1 | - | 7 - 8 | 1127 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh3a1
Genomic Map