Assay Details
| Gene Symbol: Aldh3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK051219.1 | - | 6 - 7 | 1087 | 78 | |
| AK078243.1 | - | 1087 | 78 | |||
| AK079571.1 | - | 6 - 7 | 1069 | 78 | ||
| AK079639.1 | - | 6 - 7 | 1068 | 78 | ||
| AK086262.1 | - | 6 - 7 | 1069 | 78 | ||
| AK140932.1 | - | 6 - 7 | 1107 | 78 | ||
| AK148462.1 | - | 6 - 7 | 1069 | 78 | ||
| AK152156.1 | - | 6 - 7 | 1105 | 78 | ||
| AK152735.1 | - | 6 - 7 | 1105 | 78 | ||
| AK152800.1 | - | 6 - 7 | 1105 | 78 | ||
| AK159246.1 | - | 6 - 7 | 1127 | 78 | ||
| AK163040.1 | - | 6 - 7 | 1068 | 78 | ||
| AK169157.1 | - | 6 - 7 | 1068 | 78 | ||
| AK170195.1 | - | 6 - 7 | 1088 | 78 | ||
| AK170251.1 | - | 5 - 6 | 2886 | 78 | ||
| BC003797.1 | - | 6 - 7 | 1029 | 3490519 | 78 | |
| U14390.1 | - | 6 - 7 | 1059 | 78 | ||
More Information
Set Membership: |
|

Aldh3a2
Genomic Map