Assay Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009164.2 | NP_033190.2 | 12 - 13 | 1230 | 88 | |
| GenBank mRNA | AK161215.1 | - | 11 - 12 | 1154 | 88 | |
| AK166337.1 | - | 12 - 13 | 1223 | 88 | ||
| AK172060.1 | - | 12 - 13 | 1228 | 88 | ||
| BC004598.1 | - | 10 - 11 | 951 | 3584958 | 88 | |
| X87671.1 | - | 9 - 10 | 1089 | 88 | ||
More Information
Set Membership: |
|

Sh3bp1
Genomic Map