Assay Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009164.2 | NP_033190.2 | 9 - 10 | 898 | 57 | |
| GenBank mRNA | AK145156.1 | - | 9 - 10 | 838 | 57 | |
| AK161215.1 | - | 8 - 9 | 822 | 57 | ||
| AK166337.1 | - | 9 - 10 | 891 | 57 | ||
| AK172060.1 | - | 9 - 10 | 58 | |||
| BC004598.1 | - | 7 - 8 | 3584958 | 57 | ||
| X87671.1 | - | 6 - 7 | 57 | |||
More Information
Set Membership: |
|

Sh3bp1
Genomic Map