Assay Details
| Gene Symbol: Chrnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009601.4 | NP_033731.3 | 7 - 8 | 881 | 77 | |
| GenBank mRNA | AK087554.1 | - | 7 - 8 | 860 | 77 | |
| BC023318.1 | - | 7 - 8 | 887 | 4009493 | 77 | |
| M14537.1 | - | 7 - 8 | 824 | 77 | ||
More Information
Set Membership: |
|

Chrnb1
Genomic Map