Assay Details
| Gene Symbol: Mpdu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301710.1 | NP_001288639.1 | 1 - 2 | 177 | 64 | |
| NM_001301711.1 | NP_001288640.1 | 1 - 2 | 177 | 64 | ||
| NM_011900.4 | NP_036030.2 | 1 - 2 | 177 | 64 | ||
| NR_125917.1 | 1 - 2 | 177 | 64 | |||
| GenBank mRNA | AB024713.1 | - | 1 - 2 | 105 | 64 | |
| AK040432.1 | - | 1 - 2 | 116 | 64 | ||
| AK080233.1 | - | 1 - 2 | 171 | 64 | ||
| AK089426.1 | - | 1 - 2 | 119 | 64 | ||
| AK172020.1 | - | 1 - 2 | 103 | 64 | ||
| BC026776.1 | - | 1 - 2 | 99 | 4159355 | 64 | |
| BC048871.1 | - | 1 - 2 | 105 | 5364408 | 64 | |
| CV561188.1 | - | 1 - 2 | 128 | 64 | ||
| CV561394.1 | - | 1 - 2 | 97 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001301711.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27896369 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27896371 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162452 | BC026776.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029630 | BC026776.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029630 | BC048871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029630 | AB024713.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029630 | AK040432.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2029630 | AK080233.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125219 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452615 | AK008441.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452615 | NM_001081137.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74002 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162452 | BC026776.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029630 | BC026776.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029630 | BC048871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029630 | AB024713.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029630 | AK040432.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2029630 | AK080233.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27896345 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02355938 | - | - |
| Whole Mouse Genome G4122A | A_51_P201730 | BC026776.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201730 | BC048871.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201730 | AB024713.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201730 | AK080233.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201730 | AK089426.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117898 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09455688 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09455686 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09455682 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09214815 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09214825 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27896371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27896369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27896336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09214817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09455686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09288245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09411452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09229996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27896348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09411459 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10387535 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387527 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387533 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387529 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387530 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387526 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387534 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387532 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387528 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387531 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416104_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10387525 | - | - |
| Mouse Genome 430 2.0 Array | 1416104_at | - | - |
| Mouse Genome 430 2.0 Array | 1416104_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 24070_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 24070_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100027911.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027911.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027919.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027919.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027924.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027924.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027902.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027902.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027904.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027904.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027909.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027909.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027901.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027901.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027922.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027922.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027903.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027903.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027906.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027906.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027908.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027908.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027913.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027913.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027920.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027920.mm.1 | TC1100003054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027929.mm.1 | NM_011900.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027929.mm.1 | TC1100003054.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416104_at | - | - |
| Mouse Expression Set 430 | 1416104_PM_at | - | - |
| Murine Genome U74v2 Set | 160805_s_at | - | - |
| Murine Genome U74v2 Set | 101901_at | - | - |
| Murine Genome U74v2 Set | 101901_PM_at | - | - |
| Murine Genome U74v2 Set | 160805_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416104_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416104_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416104_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416104_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 325918 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1850091 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001850091 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2685811 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2685811 | - | - |
| mouse-6 v1.1 (Illumina) | scl40021.5.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1850091 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40021.5.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1850091 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2685811 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2685811 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40021.5.1_30-S | - | - |

Mpdu1
Genomic Map