Assay Details
| Gene Symbol: Mpdu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301710.1 | NP_001288639.1 | 2 - 3 | 247 | 89 | |
| NM_001301711.1 | NP_001288640.1 | 2 - 3 | 247 | 89 | ||
| NM_011900.4 | NP_036030.2 | 2 - 3 | 247 | 89 | ||
| NR_125917.1 | 2 - 3 | 247 | 89 | |||
| GenBank mRNA | AB024713.1 | - | 2 - 3 | 175 | 89 | |
| AK040432.1 | - | 2 - 3 | 186 | 89 | ||
| AK080233.1 | - | 2 - 3 | 241 | 89 | ||
| AK089426.1 | - | 2 - 3 | 189 | 89 | ||
| AK172020.1 | - | 2 - 3 | 173 | 89 | ||
| BC026776.1 | - | 2 - 3 | 169 | 4159355 | 89 | |
| BC048871.1 | - | 2 - 3 | 175 | 5364408 | 89 | |
| CV561188.1 | - | 2 - 3 | 198 | 89 | ||
| CV561394.1 | - | 2 - 3 | 167 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001301711.1 transcripts.
Set Membership: |
|

Mpdu1
Genomic Map