Assay Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017407.2 | NP_059103.1 | 10 - 11 | 2043 | 63 | |
| GenBank mRNA | AK052228.1 | - | 9 - 10 | 2295 | 63 | |
| AK076619.1 | - | 10 - 11 | 2041 | 63 | ||
| BC026715.1 | - | 8 - 9 | 1502 | 63 | ||
| BC052672.1 | - | 10 - 11 | 2007 | 30058496 | 63 | |
| X81633.1 | - | 8 - 9 | 2237 | 63 | ||
More Information
Set Membership: |
|

Spag5
Genomic Map