Assay Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017407.2 | NP_059103.1 | 12 - 13 | 2292 | 77 | |
| GenBank mRNA | AF420307.1 | - | 12 - 13 | 2229 | 77 | |
| AK076619.1 | - | 12 - 13 | 2290 | 77 | ||
| BC026715.1 | - | 10 - 11 | 1751 | 77 | ||
| BC052672.1 | - | 12 - 13 | 2256 | 30058496 | 77 | |
More Information
Set Membership: |
|

Spag5
Genomic Map