Assay Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017407.2 | NP_059103.1 | 16 - 17 | 2785 | 68 | |
| GenBank mRNA | AF420307.1 | - | 16 - 17 | 2722 | 68 | |
| AK076619.1 | - | 16 - 17 | 2783 | 68 | ||
| BC026715.1 | - | 14 - 15 | 2244 | 68 | ||
| BC052672.1 | - | 16 - 17 | 2749 | 30058496 | 68 | |
More Information
Set Membership: |
|

Spag5
Genomic Map