Assay Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017407.2 | NP_059103.1 | 1 - 2 | 124 | 100 | |
| GenBank mRNA | AF420307.1 | - | 1 - 2 | 61 | 100 | |
| AK052228.1 | - | 1 - 2 | 92 | 100 | ||
| AK076619.1 | - | 1 - 2 | 120 | 100 | ||
| BC052672.1 | - | 1 - 2 | 88 | 30058496 | 100 | |
| X81633.1 | - | 1 - 2 | 60 | 100 | ||
More Information
Set Membership: |
|

Spag5
Genomic Map