Assay Details
| Gene Symbol: Plscr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168497.1 | NP_001161969.1 | 5 - 6 | 826 | 70 | |
| NM_023564.4 | NP_076053.1 | 5 - 6 | 750 | 70 | ||
| NR_033133.1 | 4 - 5 | 939 | 70 | |||
| GenBank mRNA | AF159850.1 | - | 4 - 5 | 688 | 70 | |
| AK011716.1 | - | 5 - 6 | 748 | 70 | ||
| AK029154.1 | - | 4 - 5 | 906 | 70 | ||
| AK075841.1 | - | 5 - 6 | 827 | 70 | ||
| AK166288.1 | - | 5 - 6 | 733 | 70 | ||
| AK172635.1 | - | 5 - 6 | 727 | 70 | ||
| BC020143.1 | - | 4 - 5 | 685 | 3967872 | 70 | |
More Information
Set Membership: |
|

Plscr3
Genomic Map