Assay Details
| Gene Symbol: Fancl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277273.1 | NP_001264202.1 | 12 - 13 | 1142 | 73 | |
| NM_025923.3 | NP_080199.1 | 12 - 13 | 1157 | 73 | ||
| NR_102382.1 | 10 - 11 | 924 | 73 | |||
| GenBank mRNA | AF513619.1 | - | 12 - 13 | 1157 | 73 | |
| AK008605.1 | - | 12 - 13 | 1043 | 73 | ||
| AK011520.1 | - | 12 - 13 | 1062 | 73 | ||
| AK011752.1 | - | 12 - 13 | 1034 | 73 | ||
| AK011903.1 | - | 12 - 13 | 1025 | 73 | ||
| AK017431.1 | - | 12 - 13 | 1037 | 73 | ||
| AK018018.1 | - | 12 - 13 | 1039 | 73 | ||
| AK076017.1 | - | 12 - 13 | 1040 | 73 | ||
| AK168538.1 | - | 12 - 13 | 1041 | 73 | ||
| BC032876.1 | - | 12 - 13 | 1031 | 1282316 | 73 | |
| BC066181.1 | - | 12 - 13 | 1086 | 6435171 | 73 | |
| CN455644.1 | - | 9 - 10 | 710 | 73 | ||
More Information
Set Membership: |
|

Fancl
Genomic Map