Assay Details
| Gene Symbol: Srfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026040.3 | NP_080316.3 | 6 - 7 | 1131 | 134 | |
| GenBank mRNA | AK012865.1 | - | 6 - 7 | 1133 | 134 | |
| AK020469.1 | - | 83 | 134 | |||
| AK141882.1 | - | 1125 | 134 | |||
| AK144845.1 | - | 1130 | 134 | |||
| AK147692.1 | - | 1109 | 134 | |||
| AY611629.1 | - | 6 - 7 | 1104 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srfbp1
Genomic Map