Assay Details
Gene Symbol: Coa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026618.2 | NP_080894.1 | 1 - 2 | 234 | 87 | |
GenBank mRNA | AK019025.1 | - | 1 - 2 | 236 | 87 | |
BC026206.1 | - | 1 - 2 | 228 | 1265058 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226364 | AK003955.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226364 | AK019025.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226364 | BC026206.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226364 | NM_026618.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226364 | AK003955.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226364 | AK019025.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226364 | BC026206.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P226364 | NM_026618.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226364 | AK003955.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226364 | AK019025.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226364 | BC026206.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226364 | NM_026618.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226364 | AK003955.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226364 | AK019025.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226364 | BC026206.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P226364 | NM_026618.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073046 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073043 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073047 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073037 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073039 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073038 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073045 | - | - |
Whole Mouse Genome G4122A | A_51_P226364 | AK003955.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P226364 | AK019025.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P226364 | BC026206.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P226364 | NM_026618.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P226364 | AK003955.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P226364 | AK019025.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P226364 | BC026206.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P226364 | NM_026618.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129368 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09208567 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09208569 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09208578 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09208573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28072970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28072973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28072967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28072971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28072966 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073043 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09208576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073040 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073037 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28073047 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10391409 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391408 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1423840_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419812_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10391407 | - | - |
Mouse Genome 430 2.0 Array | 1423840_at | - | - |
Mouse Genome 430 2.0 Array | 1419812_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1419812_s_at | - | - |
Mouse Genome 430 2.0 Array | 1423840_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52469_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52469_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100035133.mm.1 | NM_026618.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035133.mm.1 | NM_026618.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035133.mm.1 | TC1100003799.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100035131.mm.1 | NM_026618.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035131.mm.1 | NM_026618.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035131.mm.1 | TC1100003799.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100035134.mm.1 | NM_026618.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035134.mm.1 | NM_026618.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035134.mm.1 | TC1100003799.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100035132.mm.1 | NM_026618.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035132.mm.1 | NM_026618.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100035132.mm.1 | TC1100003799.mm.1 | MTA TC |
Mouse Expression Set 430 | 1419812_PM_s_at | - | - |
Mouse Expression Set 430 | 1419812_s_at | - | - |
Murine Genome U74v2 Set | 100527_at | - | - |
Murine Genome U74v2 Set | 100527_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419812_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1419812_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423840_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419812_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423840_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419812_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 644434 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1090133 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001090133 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2765287 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2765287 | - | - |
mouse-6 v1.1 (Illumina) | scl052469.1_143-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1090133 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl052469.1_143-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1090133 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2932518 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2765287 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2932518 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl052469.1_143-S | - | - |
