Assay Details
| Gene Symbol: Krt25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133730.1 | NP_598491.1 | 6 - 7 | 1212 | 89 | |
| GenBank mRNA | AK014538.1 | - | 6 - 7 | 1222 | 89 | |
| BC018391.1 | - | 6 - 7 | 1212 | 4191565 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441898 | NM_133730.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441898 | AK014538.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441898 | BC018391.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441898 | NM_133730.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441898 | AK014538.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441898 | BC018391.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676187 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 173193 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178262 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441898 | NM_133730.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441898 | AK014538.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441898 | BC018391.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441898 | NM_133730.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441898 | AK014538.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441898 | BC018391.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28061675 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178262 | - | - |
| Whole Mouse Genome G4122A | A_51_P441898 | NM_133730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P441898 | AK014538.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P441898 | BC018391.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P441898 | NM_133730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P441898 | AK014538.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P441898 | BC018391.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135754 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178262 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09191524 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09191523 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09191525 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09191531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09191528 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09191520 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09191517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28061714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09076123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09191522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09076107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09697864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09631610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09427185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09615905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09368627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28061672 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10390801 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390803 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390799 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390804 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390800 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390805 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10390802 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418173_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10390798 | - | - |
| Mouse Genome 430 2.0 Array | 1418173_at | - | - |
| Mouse Genome 430 2.0 Array | 1418173_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70810_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70810_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100034153.mm.1 | NM_133730.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034153.mm.1 | NM_133730.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034153.mm.1 | TC1100003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034155.mm.1 | NM_133730.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034155.mm.1 | NM_133730.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034155.mm.1 | TC1100003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034152.mm.1 | NM_133730.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034152.mm.1 | NM_133730.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034152.mm.1 | TC1100003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034151.mm.1 | NM_133730.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034151.mm.1 | NM_133730.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034151.mm.1 | TC1100003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034150.mm.1 | NM_133730.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034150.mm.1 | NM_133730.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034150.mm.1 | TC1100003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034154.mm.1 | NM_133730.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034154.mm.1 | NM_133730.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034154.mm.1 | TC1100003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034156.mm.1 | NM_133730.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034156.mm.1 | NM_133730.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034156.mm.1 | TC1100003702.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100034149.mm.1 | NM_133730.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034149.mm.1 | NM_133730.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100034149.mm.1 | TC1100003702.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418173_PM_at | - | - |
| Mouse Expression Set 430 | 1418173_at | - | - |
| Murine Genome U74v2 Set | 93801_at | - | - |
| Murine Genome U74v2 Set | 93801_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418173_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418173_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418173_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418173_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 642378 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5570711 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5570711 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005570711 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216096 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216096 | - | - |
| mouse-6 v1.1 (Illumina) | scl39605.8.1_106-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5570711 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39605.8.1_106-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570711 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1216096 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1216096 | - | - |

Krt25
Genomic Map