Assay Details
| Gene Symbol: Flcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271356.1 | NP_001258285.1 | 6 - 7 | 1256 | 77 | |
| NM_001271357.1 | NP_001258286.1 | 6 - 7 | 999 | 77 | ||
| NM_146018.2 | NP_666130.1 | 6 - 7 | 1296 | 77 | ||
| NR_073164.1 | 6 - 7 | 1256 | 77 | |||
| GenBank mRNA | AK039106.1 | - | 6 - 7 | 1083 | 77 | |
| AK080933.1 | - | 6 - 7 | 1081 | 77 | ||
| AK136071.1 | - | 6 - 7 | 987 | 77 | ||
| AK148216.1 | - | 6 - 7 | 1050 | 77 | ||
| AK154040.1 | - | 6 - 7 | 1278 | 77 | ||
| AK161460.1 | - | 6 - 7 | 1028 | 77 | ||
| AK172245.1 | - | 6 - 7 | 1055 | 77 | ||
| BC025820.1 | - | 6 - 7 | 1030 | 5099493 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Flcn
Genomic Map