Assay Details
| Gene Symbol: Idi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177197.4 | NP_796171.1 | 3 - 4 | 287 | 73 | |
| GenBank mRNA | AK029364.1 | - | 3 - 4 | 288 | 73 | |
| AK081319.1 | - | 3 - 4 | 288 | 73 | ||
| AY263178.1 | - | 3 - 4 | 288 | 73 | ||
| BC099586.1 | - | 3 - 4 | 274 | 30292324 | 73 | |
| BC125310.1 | - | 3 - 4 | 273 | 40129825 | 73 | |
| BC138100.1 | - | 3 - 4 | 273 | 8861119 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121137 | BC099586.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121137 | NM_177197.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121137 | AK029364.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058899 | BC125310.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058899 | BC138100.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058899 | BC099586.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058899 | AK081319.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058899 | NM_177197.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970082 | BC125310.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970082 | BC138100.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970082 | AY263178.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970082 | BC099586.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970082 | AK081319.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80753 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110143 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058899 | BC125310.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058899 | BC138100.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058899 | BC099586.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058899 | AK081319.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058899 | NM_177197.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970082 | BC125310.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970082 | BC138100.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970082 | AY263178.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970082 | BC099586.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970082 | AK081319.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121137 | BC099586.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121137 | NM_177197.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121137 | AK029364.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110143 | - | - |
| Whole Mouse Genome G4122A | A_51_P385892 | AK029364.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107540 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135624 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121137 | BC099586.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121137 | NM_177197.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121137 | AK029364.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110143 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10823023 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10841851 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10841849 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10841848 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10841854 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10841855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28813746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28813734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28813732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28813733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28813731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28813736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28813737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28813730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28813747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28813731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28813730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28813679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28813729 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10403427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407538 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407542 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407539 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407540 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407541 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403425 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440852_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10407537 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403420 | - | - |
| Mouse Genome 430 2.0 Array | 1440852_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440852_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320581_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320581_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300000329.mm.1 | NM_177197.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000329.mm.1 | NM_177197.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000329.mm.1 | TC1300000048.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300000327.mm.1 | NM_177197.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000327.mm.1 | NM_177197.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000327.mm.1 | TC1300000048.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300000328.mm.1 | NM_177197.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000328.mm.1 | NM_177197.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000328.mm.1 | TC1300000048.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300000330.mm.1 | NM_177197.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000330.mm.1 | NM_177197.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000330.mm.1 | TC1300000048.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300000332.mm.1 | NM_177197.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000332.mm.1 | NM_177197.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000332.mm.1 | TC1300000048.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300000326.mm.1 | NM_177197.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000326.mm.1 | NM_177197.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300000326.mm.1 | TC1300000048.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440852_PM_at | - | - |
| Mouse Expression Set 430 | 1440852_at | - | - |
| Murine Genome U74v2 Set | 166230_PM_at | - | - |
| Murine Genome U74v2 Set | 166230_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 627235 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3990333 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3990333 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 770035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000770035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003990333 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2689488 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216803 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216803 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2689488 | - | - |
| mouse-6 v1.1 (Illumina) | scl0320581.5_146-S | - | - |
| mouse-6 v1.1 (Illumina) | scl44305.5.1_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 770035 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3990333 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0320581.5_146-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44305.5.1_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3990333 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1216803 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2894900 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2689488 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2689488 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1216803 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2894900 | - | - |

Idi2
Genomic Map