Assay Details
| Gene Symbol: Itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010584.3 | NP_034714.1 | 8 | 922 | 119 | |
| GenBank mRNA | AB016496.1 | - | 8 | 119 | ||
| AK007447.1 | - | 8 | 922 | 119 | ||
| AY157363.1 | - | 8 | 119 | |||
| AY157364.1 | - | 8 | 906 | 119 | ||
| AY619693.1 | - | 8 | 905 | 119 | ||
| BC150795.1 | - | 8 | 864 | 9087706 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P119869 | AY157363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P119869 | AY157364.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P119869 | NM_010584.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P119869 | AB016496.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035117 | BC145376.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035117 | BC145377.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035117 | BC150760.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976022 | AY157363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976022 | AY157364.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976022 | AY619693.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976022 | NM_010584.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976022 | AB016496.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416379 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471945 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 7357 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103157 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177084 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035117 | BC145376.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035117 | BC145377.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035117 | BC150760.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976022 | AY157363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976022 | AY157364.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976022 | AY619693.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976022 | NM_010584.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976022 | AB016496.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P119869 | AY157363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P119869 | AY157364.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P119869 | NM_010584.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P119869 | AB016496.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P147987 | AY157363.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P147987 | AB016496.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20828360 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04354581 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103157 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177084 | - | - |
| Whole Mouse Genome G4122A | A_51_P147987 | AY157363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147987 | AB016496.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119869 | AY157363.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119869 | AY157364.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119869 | NM_010584.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119869 | AB016496.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103157 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177084 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00851552 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00851556 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00851555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20828347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00705949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00475586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00262013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00072254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00475585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00674680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00890895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00796612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00890900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00824395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00890901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00072256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20828374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00072251 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360157 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360151 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360152 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360156 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418165_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10360149 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351640 | - | - |
| Mouse Genome 430 2.0 Array | 1418165_at | - | - |
| Mouse Genome 430 2.0 Array | 1418165_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16429_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16429_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100029110.mm.1 | NM_010584.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029110.mm.1 | NM_010584.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029110.mm.1 | TC0100003546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100029112.mm.1 | NM_010584.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029112.mm.1 | NM_010584.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029112.mm.1 | TC0100003546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100029109.mm.1 | NM_010584.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029109.mm.1 | NM_010584.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029109.mm.1 | TC0100003546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100029114.mm.1 | NM_010584.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029114.mm.1 | NM_010584.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029114.mm.1 | TC0100003546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100029113.mm.1 | NM_010584.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029113.mm.1 | NM_010584.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029113.mm.1 | TC0100003546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100029107.mm.1 | NM_010584.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029107.mm.1 | NM_010584.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029107.mm.1 | TC0100003546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100029120.mm.1 | NM_010584.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029120.mm.1 | NM_010584.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029120.mm.1 | TC0100003546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100029111.mm.1 | NM_010584.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029111.mm.1 | NM_010584.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029111.mm.1 | TC0100003546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100029116.mm.1 | NM_010584.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029116.mm.1 | NM_010584.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029116.mm.1 | TC0100003546.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100029117.mm.1 | NM_010584.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029117.mm.1 | NM_010584.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029117.mm.1 | TC0100003546.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418165_at | - | - |
| Mouse Expression Set 430 | 1418165_PM_at | - | - |
| Murine Genome U74v2 Set | 102755_at | - | - |
| Murine Genome U74v2 Set | 102755_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418165_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418165_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418165_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418165_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 805090 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6860133 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006860133 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2772117 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2772117 | - | - |
| mouse-6 v1.1 (Illumina) | scl15934.8.1_70-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6860133 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl15934.8.1_70-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2772117 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2772117 | - | - |

Itln1
Genomic Map