Assay Details
| Gene Symbol: 1700034I23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045380.1 | 1 | 1948 | 116 | ||
| GenBank mRNA | AK006596.1 | - | 1 | 1949 | 116 | |
| AK077003.1 | - | 1 | 1907 | 116 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257841 | AK006596.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257841 | AK077003.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257841 | NM_028494.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257841 | AK006596.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257841 | AK077003.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257841 | NM_028494.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257841 | AK006596.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257841 | AK077003.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257841 | NM_028494.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257841 | AK006596.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257841 | AK077003.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257841 | NM_028494.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22082084 | - | - |
| Whole Mouse Genome G4122A | A_51_P257841 | AK006596.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257841 | AK077003.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257841 | NM_028494.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257841 | AK006596.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257841 | AK077003.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257841 | NM_028494.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112470 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02730550 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03197409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22082080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02730551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02730552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03197409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02412529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02508117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22082086 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10497962 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497963 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419016_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10497958 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10497960 | - | - |
| Mouse Genome 430 2.0 Array | 1419016_at | - | - |
| Mouse Genome 430 2.0 Array | 1419016_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73297_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73297_PM_at | - | - |
| Mouse Expression Set 430 | 1419016_at | - | - |
| Mouse Expression Set 430 | 1419016_PM_at | - | - |
| Murine Genome U74v2 Set | 166024_f_at | - | - |
| Murine Genome U74v2 Set | 166024_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419016_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419016_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419016_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419016_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 770614 | - | - |
| ABI Mouse Genome Survey Microarray | 860697 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2850021 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2350433 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002350433 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002850021 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239150 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235065 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235065 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239150 | - | - |
| mouse-6 v1.1 (Illumina) | scl073297.1_1-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0073297.1_101-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2350433 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2850021 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl073297.1_1-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0073297.1_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2350433 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850021 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2862353 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239150 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239150 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2862353 | - | - |

1700034I23Rik
Genomic Map