Assay Details
| Gene Symbol: Olfr279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001807.1 | NP_001001807.1 | 1 | 820 | 150 | |
| GenBank mRNA | BC106968.1 | - | 1 | 820 | 150 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045337 | BC106968.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045337 | NM_001001807.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130905 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045337 | BC106968.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045337 | NM_001001807.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P287956 | BC106968.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P287956 | NM_001001807.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P23366 | BC106968.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P23366 | NM_001001807.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474861 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130905 | - | - |
| Whole Mouse Genome G4122A | A_52_P23366 | BC106968.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P23366 | NM_001001807.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P287956 | BC106968.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P287956 | NM_001001807.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119629 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130905 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11845694 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11845702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12274169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12262650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11878076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12274172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12177268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11845688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11912440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474826 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10426602 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10426601 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 478391 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3163554 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3163554 | - | - |
| mouse-6 v1.1 (Illumina) | GI_49227366-A | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3163554 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3068120 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3163554 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3068120 | - | - |

Olfr279
Genomic Map